Anti-SPG20 Goat Antibody

Lieferant: OriGene
TA311430
ORIGTA311430EA 785 EUR
ORIGTA311430
Anti-SPG20 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml; Immunofluorescence: 2?g/ml

Type: Primary
Antigen: SPG20 (spastic paraplegia 20 (Troyer syndrome))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR