Anti-RSL1D1 Goat Antibody

Lieferant: OriGene
TA311444
ORIGTA311444EA 785 EUR
ORIGTA311444
Anti-RSL1D1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: RSL1D1 (ribosomal L1 domain containing 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR