Anti-PI15 Goat Antibody

Lieferant: OriGene
TA311447
ORIGTA311447EA 785 EUR
ORIGTA311447
Anti-PI15 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: PI15 (peptidase inhibitor 15)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR