Anti-GAL Goat Antibody

Lieferant: OriGene
TA311451
ORIGTA311451EA 785 EUR
ORIGTA311451
Anti-GAL Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml; Immunochemistry: 0.2-0.5?g/ml

Type: Primary
Antigen: GAL (galanin/GMAP prepropeptide)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR