Anti-RHCG Goat Antibody

Lieferant: OriGene
TA311456
ORIGTA311456EA 785 EUR
ORIGTA311456
Anti-RHCG Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: RHCG (Rh family, C glycoprotein)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR