Anti-CCAR1 Goat Antibody

Lieferant: OriGene
TA311476
ORIGTA311476EA 785 EUR
ORIGTA311476
Anti-CCAR1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: CCAR1 (cell division cycle and apoptosis regulator 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR