Anti-CYP26B1 Goat Antibody

Lieferant: OriGene
TA311484
ORIGTA311484EA 785 EUR
ORIGTA311484
Anti-CYP26B1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.1?g/ml; Immunochemistry: 4-6?g/ml

Type: Primary
Antigen: CYP26B1 (cytochrome P450, family 26, subfamily B, polypeptide 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR