Anti-PRKCQ Rabbit Polyclonal Antibody

Lieferant: OriGene
TA312813
ORIGTA312813EA 528 EUR
ORIGTA312813
Anti-PRKCQ Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: PRKCQ (protein kinase C, theta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR