Anti-KRT8 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA313114
ORIGTA313114EA 528 EUR
ORIGTA313114
Anti-KRT8 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-3000; Immunochemistry: 1:50-100; ELISA: 1:40000

Type: Primary
Antigen: KRT8 (keratin 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR