Anti-ATP5A1 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA313487
ORIGTA313487EA 528 EUR
ORIGTA313487
Anti-ATP5A1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: ATP5A1 (ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1, cardiac muscle)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR