Anti-CYP2C18 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA313723
ORIGTA313723EA 528 EUR
ORIGTA313723
Anti-CYP2C18 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: CYP2C18 (cytochrome P450, family 2, subfamily C, polypeptide 18)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR