Anti-EEF1G Rabbit Polyclonal Antibody

Lieferant: OriGene
TA313785
ORIGTA313785EA 528 EUR
ORIGTA313785
Anti-EEF1G Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: EEF1G (eukaryotic translation elongation factor 1 gamma)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR