Anti-F2RL2 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA313856
ORIGTA313856EA 528 EUR
ORIGTA313856
Anti-F2RL2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: F2RL2 (coagulation factor II (thrombin) receptor-like 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR