Anti-PPP1R2 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA314371
ORIGTA314371EA 528 EUR
ORIGTA314371
Anti-PPP1R2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: PPP1R2 (protein phosphatase 1, regulatory (inhibitor) subunit 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR