Anti-KLF7 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA314780
ORIGTA314780EA 528 EUR
ORIGTA314780
Anti-KLF7 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: KLF7 (Kruppel-like factor 7 (ubiquitous))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR