Anti-ACTR3 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA315015
ORIGTA315015EA 528 EUR
ORIGTA315015
Anti-ACTR3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: ACTR3 (ARP3 actin-related protein 3 homolog (yeast))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR