Anti-TOB1 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA315019
ORIGTA315019EA 528 EUR
ORIGTA315019
Anti-TOB1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: TOB1 (transducer of ERBB2, 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR