Anti-CDK2AP2 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA315040
ORIGTA315040EA 528 EUR
ORIGTA315040
Anti-CDK2AP2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:1000

Type: Primary
Antigen: CDK2AP2 (cyclin-dependent kinase 2 associated protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR