Anti-CLIP3 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA315300
ORIGTA315300EA 528 EUR
ORIGTA315300
Anti-CLIP3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: CLIP3 (CAP-GLY domain containing linker protein 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR