Anti-OR4D1 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA315345
ORIGTA315345EA 528 EUR
ORIGTA315345
Anti-OR4D1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: OR4D1 (olfactory receptor, family 4, subfamily D, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR