Anti-MRPL4 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA315442
ORIGTA315442EA 528 EUR
ORIGTA315442
Anti-MRPL4 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: MRPL4 (mitochondrial ribosomal protein L4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR