Anti-RFWD2 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA315725
ORIGTA315725EA 528 EUR
ORIGTA315725
Anti-RFWD2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: RFWD2 (ring finger and WD repeat domain 2, E3 ubiquitin protein ligase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR