Anti-CYP3A43 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA315747
ORIGTA315747EA 528 EUR
ORIGTA315747
Anti-CYP3A43 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: CYP3A43 (cytochrome P450, family 3, subfamily A, polypeptide 43)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR