Anti-TRAPPC6A Rabbit Polyclonal Antibody

Lieferant: OriGene
TA315773
ORIGTA315773EA 528 EUR
ORIGTA315773
Anti-TRAPPC6A Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: TRAPPC6A (trafficking protein particle complex 6A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR