Anti-OR2L5 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA315851
ORIGTA315851EA 528 EUR
ORIGTA315851
Anti-OR2L5 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: OR2L5 (olfactory receptor, family 2, subfamily L, member 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR