Anti-CDCA7 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA315887
ORIGTA315887EA 528 EUR
ORIGTA315887
Anti-CDCA7 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: CDCA7 (cell division cycle associated 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR