Anti-OR8U1 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA316226
ORIGTA316226EA 528 EUR
ORIGTA316226
Anti-OR8U1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: OR8U1 (olfactory receptor, family 8, subfamily U, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR