Anti-OR6C3 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA316271
ORIGTA316271EA 528 EUR
ORIGTA316271
Anti-OR6C3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: OR6C3 (olfactory receptor, family 6, subfamily C, member 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR