Anti-CDK1 Mouse Monoclonal Antibody [clone: 26E11]

Lieferant: OriGene
TA316617
ORIGTA316617EA 694 EUR
ORIGTA316617
Anti-CDK1 Mouse Monoclonal Antibody [clone: 26E11]
Antikörper
Recommended Dilutions: Immunochemistry: 1:200 Western Blot: 1:1000

Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: Monoclonal
Clone: 26E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR