Anti-DRD3 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA316682
ORIGTA316682EA 672 EUR
ORIGTA316682
Anti-DRD3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 20 µg/ml

Type: Primary
Antigen: DRD3 (dopamine receptor D3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR