Anti-ENPP3 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA316707
ORIGTA316707EA 715 EUR
ORIGTA316707
Anti-ENPP3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 15-20 µg/ml

Type: Primary
Antigen: ENPP3 (ectonucleotide pyrophosphatase/phosphodiesterase 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR