Anti-GPR84 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA316854
ORIGTA316854EA 715 EUR
ORIGTA316854
Anti-GPR84 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 10-13 µg/ml

Type: Primary
Antigen: GPR84 (G protein-coupled receptor 84)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR