Anti-NLRP8 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA317067
ORIGTA317067EA 743 EUR
ORIGTA317067
Anti-NLRP8 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 5 µg/ml; Western Blot: 1-2 µg/ml

Type: Primary
Antigen: NLRP8 (NLR family, pyrin domain containing 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR