Anti-OXGR1 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA317121
ORIGTA317121EA 715 EUR
ORIGTA317121
Anti-OXGR1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 22 µg/ml

Type: Primary
Antigen: OXGR1 (oxoglutarate (alpha-ketoglutarate) receptor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR