Anti-SLC2A2 Goat Polyclonal Antibody

Lieferant: OriGene
TA317322
ORIGTA317322EA 743 EUR
ORIGTA317322
Anti-SLC2A2 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 2.5 µg/ml

Type: Primary
Antigen: SLC2A2 (solute carrier family 2 (facilitated glucose transporter), member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR