Anti-SLC39A6 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA317332
ORIGTA317332EA 715 EUR
ORIGTA317332
Anti-SLC39A6 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Immunochemistry: 10 µg/ml

Type: Primary
Antigen: SLC39A6 (solute carrier family 39 (zinc transporter), member 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR