Anti-SLC5A10 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA317338
ORIGTA317338EA 714 EUR
ORIGTA317338
Anti-SLC5A10 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 20 µg/ml

Type: Primary
Antigen: SLC5A10 (solute carrier family 5 (sodium/sugar cotransporter), member 10)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR