Anti-TM9SF3 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA317428
ORIGTA317428EA 715 EUR
ORIGTA317428
Anti-TM9SF3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 10 µg/ml

Type: Primary
Antigen: TM9SF3 (transmembrane 9 superfamily member 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR