Anti-WNT10A Rabbit Polyclonal Antibody

Lieferant: OriGene
TA317690
ORIGTA317690EA 715 EUR
ORIGTA317690
Anti-WNT10A Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 10 µg/ml

Type: Primary
Antigen: WNT10A (wingless-type MMTV integration site family, member 10A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR