Anti-WNT4 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA317957
ORIGTA317957EA 715 EUR
ORIGTA317957
Anti-WNT4 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 10 µg/ml

Type: Primary
Antigen: WNT4 (wingless-type MMTV integration site family, member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR