Anti-B2M Mouse Monoclonal Antibody

Lieferant: OriGene
TA318043
ORIGTA318043EA 799 EUR
ORIGTA318043
Anti-B2M Mouse Monoclonal Antibody
Antikörper
Recommended Dilutions: ELISA Flo: 1 µg/ml; Immunochemistry: 10 µg/ml Western Blot: 2-4 µg/ml

Type: Primary
Antigen: B2M (beta-2-microglobulin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR