Anti-KCNJ6 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA318841
ORIGTA318841EA 715 EUR
ORIGTA318841
Anti-KCNJ6 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 20 µg/ml

Type: Primary
Antigen: KCNJ6 (potassium inwardly-rectifying channel, subfamily J, member 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR