Anti-PRMT6 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA318867
ORIGTA318867EA 565 EUR
ORIGTA318867
Anti-PRMT6 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: PRMT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR