Anti-Capn1 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA318876
ORIGTA318876EA 672 EUR
ORIGTA318876
Anti-Capn1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: Capn1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Pig, Rabbit, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR