Anti-Tnfrsf21 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA318878
ORIGTA318878EA 565 EUR
ORIGTA318878
Anti-Tnfrsf21 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: Tnfrsf21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR