Anti-Siah3 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA318883
ORIGTA318883EA 565 EUR
ORIGTA318883
Anti-Siah3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-4 ?g/ml

Type: Primary
Antigen: Siah3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR