Anti-Angptl4 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA318914
ORIGTA318914EA 604 EUR
ORIGTA318914
Anti-Angptl4 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: Angptl4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR