Anti-Serpinh1 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA318915
ORIGTA318915EA 604 EUR
ORIGTA318915
Anti-Serpinh1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-4 ?g/ml; Immunochemistry: 2.5?g/ml; Immunoprecipitation: 20?g/ml

Type: Primary
Antigen: Serpinh1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR