Anti-Mapk8ip1 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA318944
ORIGTA318944EA 565 EUR
ORIGTA318944
Anti-Mapk8ip1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: Mapk8ip1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR