Anti-UBA2 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA319807
ORIGTA319807EA 792 EUR
ORIGTA319807
Anti-UBA2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.25 ?g/ml; Immunochemistry: 4 ?g/ml; Immunofluorescence: 5 ?g/ml

Type: Primary
Antigen: UBA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR